siRNA Details
Gene Symbol: C1orf210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164829.1 | NP_001158301.1 | 3 | 743 | ||
NM_182517.2 | NP_872323.1 | 3 | 814 | |||
XM_011540802.1 | XP_011539104.1 | 3 | 873 | |||
GenBank mRNA | BC041633.1 | - | 3 | 803 | 4868093 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
