siRNA Details
Gene Symbol: COMMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099339.1 | NP_001092809.1 | 7 | 1072 | ||
NM_053041.2 | NP_444269.2 | 7 | 1075 | |||
XM_005260299.3 | XP_005260356.1 | 7 | 1062 | |||
XM_005260300.3 | XP_005260357.1 | 7 | 1059 | |||
XM_011528604.2 | XP_011526906.1 | 7 | 1062 | |||
XM_017027685.1 | XP_016883174.1 | 7 | 1059 | |||
GenBank mRNA | AJ012490.1 | - | 2 | 62 | ||
AK027893.1 | - | 7 | 522 | |||
AK096307.1 | - | 7 | 1064 | |||
AK097059.1 | - | 8 | 599 | |||
AY542162.1 | - | 7 | 458 | |||
BC000628.2 | - | 7 | 508 | 3343149 | ||
BC047440.1 | - | 7 | 636 | 5275167 |
