siRNA Details
Gene Symbol: LSM14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005260302.3 | XP_005260359.1 | 3, 4 | 630 | ||
XM_011528605.2 | XP_011526907.1 | 3, 4 | 714 | |||
XM_011528606.2 | XP_011526908.1 | 3, 4 | 632 | |||
XM_011528607.2 | XP_011526909.1 | 3, 4 | 718 | |||
XM_011528608.2 | XP_011526910.1 | 3, 4 | 711 | |||
XM_011528612.2 | XP_011526914.1 | 3, 4 | 384 | |||
XM_011528613.2 | XP_011526915.1 | 3, 4 | 352 | |||
XM_011528614.2 | XP_011526916.1 | 3, 4 | 565 | |||
GenBank mRNA | AK058202.1 | - | 3, 4 | 389 | ||
AK298748.1 | - | 2, 3 | 227 | |||
BC057387.1 | - | 3, 4 | 635 | 5724236 | ||
BM547301.1 | - | 3, 4 | 635 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
