siRNA Details
Gene Symbol: C22orf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028602.1 | XP_016884091.1 | 3 | 1412 | ||
XM_017028603.1 | XP_016884092.1 | 3 | 1391 | |||
XM_017028606.1 | XP_016884095.1 | 3 | 1412 | |||
XM_017028607.1 | XP_016884096.1 | 3 | 1412 | |||
XM_017028608.1 | XP_016884097.1 | 3 | 1412 | |||
XM_017028609.1 | XP_016884098.1 | 4 | 1141 | |||
XM_017028610.1 | XP_016884099.1 | 4 | 1119 | |||
GenBank mRNA | AK093880.1 | - | 3 | 878 | ||
BC113450.1 | - | 1 | 240 | 8322502 | ||
BC113452.1 | - | 1 | 240 | 8322504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
