siRNA Details
Gene Symbol: C22orf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093880.1 | - | 3 | 747 | ||
BC113450.1 | - | 1 | 109 | 8322502 | ||
BC113452.1 | - | 1 | 109 | 8322504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
