siRNA Details
Gene Symbol: LOC150935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037808.1 | 3 | 262 | |||
GenBank mRNA | AK056439.1 | - | 3 | 262 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC150935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037808.1 | 3 | 262 | |||
GenBank mRNA | AK056439.1 | - | 3 | 262 |