siRNA Details
Gene Symbol: ZNF385B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113397.1 | NP_001106868.1 | 4 | 736 | ||
NM_001113398.1 | NP_001106869.1 | 4 | 596 | |||
NM_001282725.1 | NP_001269654.1 | 6 | 690 | |||
NM_152520.4 | NP_689733.3 | 6 | 1263 | |||
NR_104234.1 | 8 | 934 | ||||
XM_011510713.2 | XP_011509015.1 | 7 | 4323 | |||
XM_011510714.2 | XP_011509016.1 | 6 | 4273 | |||
XM_011510715.2 | XP_011509017.1 | 7 | 4307 | |||
XM_011510716.2 | XP_011509018.1 | 6 | 1434 | |||
XM_011510717.2 | XP_011509019.1 | 6 | 1431 | |||
XM_011510719.2 | XP_011509021.1 | 6 | 694 | |||
XM_011510720.2 | XP_011509022.1 | 5 | 554 | |||
XM_011510721.2 | XP_011509023.1 | 4 | 2847 | |||
XM_011510723.2 | XP_011509025.1 | 7 | 4347 | |||
XM_017003435.1 | XP_016858924.1 | 6 | 944 | |||
XM_017003436.1 | XP_016858925.1 | 5 | 1341 | |||
XM_017003437.1 | XP_016858926.1 | 5 | 579 | |||
XM_017003438.1 | XP_016858927.1 | 4 | 6062 | |||
XM_017003439.1 | XP_016858928.1 | 4 | 570 | |||
XM_017003440.1 | XP_016858929.1 | 5 | 4099 | |||
XM_017003441.1 | XP_016858930.1 | 6 | 4300 | |||
GenBank mRNA | AK057999.1 | - | 4 | 596 | ||
AK096822.1 | - | 4 | 736 | |||
BC038422.1 | - | 6 | 688 | 5165975 | ||
BC048123.1 | - | 8 | 937 | |||
BC092423.1 | - | 6 | 1098 | 30378128 |
