siRNA Details
Gene Symbol: ZNF385B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113397.1 | NP_001106868.1 | 4 | 607 | ||
NM_001113398.1 | NP_001106869.1 | 4 | 467 | |||
NM_001282725.1 | NP_001269654.1 | 6 | 561 | |||
NM_152520.4 | NP_689733.3 | 6 | 1134 | |||
NR_104234.1 | 8 | 805 | ||||
XM_011510713.2 | XP_011509015.1 | 7 | 4194 | |||
XM_011510714.2 | XP_011509016.1 | 6 | 4144 | |||
XM_011510715.2 | XP_011509017.1 | 7 | 4178 | |||
XM_011510716.2 | XP_011509018.1 | 6 | 1305 | |||
XM_011510717.2 | XP_011509019.1 | 6 | 1302 | |||
XM_011510719.2 | XP_011509021.1 | 6 | 565 | |||
XM_011510720.2 | XP_011509022.1 | 5 | 425 | |||
XM_011510721.2 | XP_011509023.1 | 4 | 2718 | |||
XM_011510723.2 | XP_011509025.1 | 7 | 4218 | |||
XM_017003435.1 | XP_016858924.1 | 6 | 815 | |||
XM_017003436.1 | XP_016858925.1 | 5 | 1212 | |||
XM_017003437.1 | XP_016858926.1 | 5 | 450 | |||
XM_017003438.1 | XP_016858927.1 | 4 | 5933 | |||
XM_017003439.1 | XP_016858928.1 | 4 | 441 | |||
XM_017003440.1 | XP_016858929.1 | 5 | 3970 | |||
XM_017003441.1 | XP_016858930.1 | 6 | 4171 | |||
GenBank mRNA | AK057999.1 | - | 4 | 467 | ||
AK096822.1 | - | 4 | 607 | |||
BC038422.1 | - | 6 | 559 | 5165975 | ||
BC048123.1 | - | 8 | 808 | |||
BC092423.1 | - | 6 | 969 | 30378128 |

More Information
Other Species Targeted

Gene Ontology Categories:
