siRNA Details
Gene Symbol: CCNYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152523.2 | NP_689736.1 | 7 | 612 | ||
XM_005246343.1 | XP_005246400.1 | 6 | 630 | |||
GenBank mRNA | AK095479.1 | - | 1 | 51 | ||
AK097751.1 | - | 7 | 612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
