siRNA Details
Gene Symbol: PQLC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099777.3 | NP_001093247.1 | 4 | 429 | ||
NM_001130002.2 | NP_001123474.1 | 4 | 429 | |||
XM_017005754.1 | XP_016861243.1 | 4 | 383 | |||
GenBank mRNA | BC029129.1 | - | 4 | 376 | ||
BC126187.1 | - | 4 | 396 | 8991903 | ||
BC126189.1 | - | 4 | 396 | 8991905 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
