siRNA Details
Gene Symbol: CCDC152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134848.1 | NP_001128320.1 | 2 | 97 | ||
GenBank mRNA | AK127368.1 | - | 2 | 87 | ||
AK303469.1 | - | 2 | 97 | |||
AY947526.1 | - | 1 | 28 | |||
BC015985.1 | - | 2 | 90 | |||
BC039102.1 | - | 2 | 69 | |||
DB450032.1 | - | 2 | 97 |
