siRNA Details
Gene Symbol: TSTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113205.1 | NP_001106676.1 | 3 | 655 | ||
NM_001113206.1 | NP_001106677.1 | 3 | 446 | |||
NM_001113207.1 | NP_001106678.1 | 4 | 569 | |||
GenBank mRNA | BC001699.1 | - | 3 | 601 | ||
BC038219.1 | - | 3 | 601 | |||
CD675389.1 | - | 4 | 564 |
