siRNA Details
Gene Symbol: PLEKHG4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052909.3 | NP_443141.3 | 12 | 2532 | ||
XM_006714446.3 | XP_006714509.1 | 14 | 3670 | |||
XM_011513968.2 | XP_011512270.1 | 14 | 3667 | |||
XM_011513969.2 | XP_011512271.1 | 14 | 3547 | |||
XM_011513970.1 | XP_011512272.1 | 14 | 3259 | |||
XM_011513971.2 | XP_011512273.1 | 14 | 3615 | |||
XM_017009112.1 | XP_016864601.1 | 14 | 3671 | |||
GenBank mRNA | AB067496.2 | - | 12 | 2532 | ||
AK125163.1 | - | 1 | 361 |
