siRNA Details
Gene Symbol: IRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033267.4 | NP_150366.1 | 4 | 1870 | ||
GenBank mRNA | AB188492.1 | - | 4 | 1873 | ||
AI928035.1 | - | 2 | 372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
