siRNA Details
Gene Symbol: ZNF720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130913.1 | NP_001124385.1 | 5 | 1040 | ||
GenBank mRNA | AL832197.1 | - | 5 | 5936 | ||
BC018134.1 | - | 5 | 995 | 3912539 | ||
BC055408.1 | - | 5 | 1036 | 3684126 |
