siRNA Details
Gene Symbol: C20orf194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009984.2 | NP_001009984.1 | 12 | 1196 | ||
XM_017027783.1 | XP_016883272.1 | 9 | 817 | |||
GenBank mRNA | AK226100.1 | - | 7 | 620 | ||
AK303375.1 | - | 3 | 568 | |||
AK310540.1 | - | 12 | 1214 | |||
AL110249.1 | - | 3 | 161 | |||
BC122529.1 | - | 3 | 414 | 40012183 |
