siRNA Details
Gene Symbol: TMEM251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098621.1 | NP_001092091.1 | 2 | 1175 | ||
NM_015676.1 | NP_056491.1 | 2 | 801 | |||
GenBank mRNA | AL080118.1 | - | 2 | 770 | ||
BC030119.2 | - | 2 | 768 | |||
BP398505.1 | - | 1 | 398 |
