siRNA Details
Gene Symbol: TMEM151B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137560.1 | NP_001131032.1 | 3 | 3420 | ||
GenBank mRNA | AK094365.1 | - | 1 | 1421 | ||
AK124502.1 | - | 1 | 1918 | |||
AK126839.1 | - | 2 | 2000 | |||
L35592.1 | - | 1 | 953 |
