siRNA Details
Gene Symbol: RUNDC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134405.1 | NP_001127877.1 | 9 | 1482 | ||
NM_138290.2 | NP_612147.1 | 10 | 1533 | |||
XM_005250156.2 | XP_005250213.1 | 9 | 1482 | |||
XM_005250158.2 | XP_005250215.1 | 7 | 957 | |||
XM_011515826.1 | XP_011514128.1 | 10 | 1533 | |||
XM_011515828.2 | XP_011514130.1 | 10 | 1016 | |||
XM_011515829.2 | XP_011514131.1 | 9 | 1150 | |||
XM_017011761.1 | XP_016867250.1 | 10 | 1220 | |||
XM_017011762.1 | XP_016867251.1 | 11 | 1145 | |||
XM_017011763.1 | XP_016867252.1 | 8 | 959 | |||
XM_017011764.1 | XP_016867253.1 | 7 | 957 | |||
GenBank mRNA | AK055233.1 | - | 10 | 1531 | ||
AK293607.1 | - | 9 | 1298 | |||
AK294039.1 | - | 9 | 1280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
