siRNA Details
Gene Symbol: RUNDC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134405.1 | NP_001127877.1 | 5 | 934 | ||
NM_001134406.1 | NP_001127878.1 | 5 | 934 | |||
NM_138290.2 | NP_612147.1 | 6 | 985 | |||
XM_005250156.2 | XP_005250213.1 | 5 | 934 | |||
XM_005250158.2 | XP_005250215.1 | 3 | 409 | |||
XM_011515826.1 | XP_011514128.1 | 6 | 985 | |||
XM_011515827.1 | XP_011514129.1 | 6 | 985 | |||
XM_011515828.2 | XP_011514130.1 | 6 | 468 | |||
XM_011515829.2 | XP_011514131.1 | 5 | 602 | |||
XM_017011761.1 | XP_016867250.1 | 6 | 672 | |||
XM_017011762.1 | XP_016867251.1 | 7 | 597 | |||
XM_017011763.1 | XP_016867252.1 | 4 | 411 | |||
XM_017011764.1 | XP_016867253.1 | 3 | 409 | |||
XM_017011765.1 | XP_016867254.1 | 6 | 584 | |||
XM_017011766.1 | XP_016867255.1 | 6 | 863 | |||
GenBank mRNA | AK055233.1 | - | 6 | 983 | ||
AK293607.1 | - | 5 | 750 | |||
AK294039.1 | - | 5 | 732 | |||
AY033596.1 | - | 4 | 420 | |||
BC022520.1 | - | 5 | 885 | 4793693 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
