siRNA Details
Gene Symbol: BMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152556.2 | NP_689769.2 | 5 | 1010 | ||
GenBank mRNA | AK056380.1 | - | 5 | 1009 | ||
AK299901.1 | - | 7 | 856 | |||
AL834437.1 | - | 5 | 805 | |||
BC114615.1 | - | 5 | 1002 | 40032372 |
