siRNA Details
Gene Symbol: C10orf131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130446.2 | NP_001123918.2 | 6 | 384 | ||
GenBank mRNA | AK294399.1 | - | 6 | 384 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C10orf131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130446.2 | NP_001123918.2 | 6 | 384 | ||
GenBank mRNA | AK294399.1 | - | 6 | 384 |