siRNA Details
Gene Symbol: FAM201A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027294.1 | 2 | 1424 | |||
GenBank mRNA | BC036230.1 | - | 2 | 1423 | ||
BC041970.1 | - | 2 | 1435 | |||
BC067887.1 | - | 1 | 1341 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM201A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027294.1 | 2 | 1424 | |||
GenBank mRNA | BC036230.1 | - | 2 | 1423 | ||
BC041970.1 | - | 2 | 1435 | |||
BC067887.1 | - | 1 | 1341 |