siRNA Details
Gene Symbol: MOSPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152581.3 | NP_689794.1 | 2, 3 | 201 | ||
XM_005274451.1 | XP_005274508.1 | 2, 3 | 201 | |||
GenBank mRNA | AK315781.1 | - | 2, 3 | 142 | ||
BC030641.1 | - | 2, 3 | 168 | 4821861 | ||
DA708010.1 | - | 2, 3 | 201 |
