siRNA Details
Gene Symbol: WDFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020945.1 | NP_065996.1 | 5, 6 | 671 | ||
XM_005270004.3 | XP_005270061.1 | 5, 6 | 786 | |||
XM_011539986.2 | XP_011538288.2 | 5, 6 | 789 | |||
XM_011539987.2 | XP_011538289.2 | 5, 6 | 792 | |||
XM_011539988.2 | XP_011538290.1 | 6, 7 | 870 | |||
XM_011539990.2 | XP_011538292.1 | 5, 6 | 817 | |||
XM_011539991.2 | XP_011538293.1 | 5, 6 | 821 | |||
XM_011539992.2 | XP_011538294.1 | 5, 6 | 821 | |||
XM_017016463.1 | XP_016871952.1 | 6, 7 | 713 | |||
XM_017016465.1 | XP_016871954.1 | 5, 6 | 798 | |||
GenBank mRNA | AK127650.1 | - | 5, 6 | 680 | ||
BC034937.1 | - | 5, 6 | 737 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
