siRNA Details
Gene Symbol: WDFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020945.1 | NP_065996.1 | 2 | 231 | ||
XM_005270004.3 | XP_005270061.1 | 2 | 346 | |||
XM_011539986.2 | XP_011538288.2 | 2 | 349 | |||
XM_011539987.2 | XP_011538289.2 | 2 | 352 | |||
XM_011539988.2 | XP_011538290.1 | 3 | 430 | |||
XM_011539990.2 | XP_011538292.1 | 2 | 377 | |||
XM_011539991.2 | XP_011538293.1 | 2 | 381 | |||
XM_011539992.2 | XP_011538294.1 | 2 | 381 | |||
XM_017016463.1 | XP_016871952.1 | 3 | 273 | |||
XM_017016465.1 | XP_016871954.1 | 2 | 358 | |||
GenBank mRNA | AK127650.1 | - | 2 | 240 | ||
BC034937.1 | - | 2 | 297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
