siRNA Details
Gene Symbol: ZNF100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173531.3 | NP_775802.2 | 5 | 680 | ||
GenBank mRNA | AK126551.1 | - | 5 | 630 | ||
BC031248.1 | - | 4 | 645 | |||
BC035579.1 | - | 5 | 592 | 5576923 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
