siRNA Details
Gene Symbol: PLPPR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010861.2 | NP_001010861.1 | 3 | 826 | ||
NM_001037317.1 | NP_001032394.1 | 3 | 826 | |||
XM_011540836.2 | XP_011539138.1 | 3 | 820 | |||
XM_011540837.1 | XP_011539139.1 | 3 | 820 | |||
XM_011540838.2 | XP_011539140.1 | 4 | 1301 | |||
GenBank mRNA | AK295688.1 | - | 3 | 564 | ||
BC040174.1 | - | 3 | 563 | |||
BC136350.1 | - | 3 | 764 | 9020337 | ||
BC144000.1 | - | 3 | 764 | 9052518 | ||
FJ472844.1 | - | 3 | 826 |
