siRNA Details
Gene Symbol: SPRR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173080.2 | NP_775103.1 | 2 | 217 | ||
XM_017000482.1 | XP_016855971.1 | 3 | 913 | |||
GenBank mRNA | BC069445.1 | - | 2 | 205 | 7262727 | |
BC069549.1 | - | 2 | 205 | 7262739 | ||
BC112170.1 | - | 2 | 217 | 8327638 | ||
BC112172.1 | - | 2 | 215 | 8327640 | ||
W77994.1 | - | 1 | 150 |
