siRNA Details
Gene Symbol: CNST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152609.2 | NP_689822.2 | 11 | 2430 | ||
XM_005273081.3 | XP_005273138.2 | 11 | 2215 | |||
XM_005273083.4 | XP_005273140.1 | 10 | 2133 | |||
XM_011544110.2 | XP_011542412.1 | 11 | 2281 | |||
XM_011544111.1 | XP_011542413.1 | 11 | 2452 | |||
XM_011544112.1 | XP_011542414.1 | 10 | 1999 | |||
XM_011544113.1 | XP_011542415.1 | 10 | 2124 | |||
XM_011544114.2 | XP_011542416.1 | 10 | 1839 | |||
XM_017000487.1 | XP_016855976.1 | 12 | 2470 | |||
XM_017000488.1 | XP_016855977.1 | 11 | 2213 | |||
XM_017000489.1 | XP_016855978.1 | 11 | 2280 | |||
GenBank mRNA | AK056563.1 | - | 11 | 2263 | ||
AK074142.1 | - | 11 | 2194 | |||
AK091739.1 | - | 3 | 976 | |||
AL834246.2 | - | 11 | 2264 | |||
BX647629.1 | - | 11 | 2215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
