siRNA Details
Gene Symbol: CNST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139459.1 | NP_001132931.1 | 6 | 1246 | ||
NM_152609.2 | NP_689822.2 | 6 | 1120 | |||
XM_005273081.3 | XP_005273138.2 | 6 | 905 | |||
XM_005273083.4 | XP_005273140.1 | 5 | 823 | |||
XM_011544110.2 | XP_011542412.1 | 6 | 971 | |||
XM_011544111.1 | XP_011542413.1 | 6 | 1142 | |||
XM_011544112.1 | XP_011542414.1 | 5 | 689 | |||
XM_011544113.1 | XP_011542415.1 | 5 | 814 | |||
XM_011544114.2 | XP_011542416.1 | 5 | 529 | |||
XM_017000487.1 | XP_016855976.1 | 7 | 1160 | |||
XM_017000488.1 | XP_016855977.1 | 6 | 903 | |||
XM_017000489.1 | XP_016855978.1 | 6 | 970 | |||
GenBank mRNA | AK056563.1 | - | 6 | 953 | ||
AL834246.2 | - | 6 | 954 | |||
BC010228.2 | - | 6 | 1122 | 4151228 | ||
BC036200.1 | - | 5 | 1475 | |||
BX647629.1 | - | 6 | 905 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
