siRNA Details
Gene Symbol: CNST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139459.1 | NP_001132931.1 | 3 | 903 | ||
NM_152609.2 | NP_689822.2 | 3 | 777 | |||
XM_005273081.3 | XP_005273138.2 | 3 | 562 | |||
XM_011544110.2 | XP_011542412.1 | 3 | 628 | |||
XM_011544111.1 | XP_011542413.1 | 3 | 799 | |||
XM_011544112.1 | XP_011542414.1 | 2 | 346 | |||
XM_011544114.2 | XP_011542416.1 | 2 | 236 | |||
XM_017000487.1 | XP_016855976.1 | 4 | 817 | |||
XM_017000488.1 | XP_016855977.1 | 3 | 610 | |||
XM_017000489.1 | XP_016855978.1 | 3 | 677 | |||
GenBank mRNA | AK056563.1 | - | 3 | 610 | ||
AK074142.1 | - | 3 | 540 | |||
AL834246.2 | - | 3 | 611 | |||
BC010228.2 | - | 3 | 779 | 4151228 | ||
BC036200.1 | - | 3 | 670 | |||
BX647629.1 | - | 3 | 562 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
