siRNA Details
Gene Symbol: ZNF366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152625.2 | NP_689838.1 | 5 | 2128 | ||
GenBank mRNA | AK097115.1 | - | 5 | 2128 | ||
BC121053.1 | - | 5 | 1999 | 40118323 | ||
BC121054.1 | - | 5 | 1999 | 40118325 | ||
BX647592.1 | - | 5 | 2018 |
