siRNA Details
Gene Symbol: RNF133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139175.1 | NP_631914.1 | 1 | 522 | ||
GenBank mRNA | AF447589.1 | - | 1 | 522 | ||
AK098524.1 | - | 1 | 524 | |||
BC022038.2 | - | 1 | 501 | 4823227 |
