siRNA Details
Gene Symbol: ZNF169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301275.1 | NP_001288204.1 | 5 | 2130 | ||
NM_003448.2 | NP_003439.2 | 5 | 1766 | |||
NM_194320.3 | NP_919301.2 | 5 | 1763 | |||
XM_017014362.1 | XP_016869851.1 | 5 | 1895 | |||
XM_017014363.1 | XP_016869852.1 | 6 | 1789 | |||
XM_017014364.1 | XP_016869853.1 | 6 | 1786 | |||
XM_017014365.1 | XP_016869854.1 | 5 | 8119 | |||
XM_017014366.1 | XP_016869855.1 | 5 | 8057 | |||
GenBank mRNA | AK093956.1 | - | 5 | 2068 | ||
AK122832.1 | - | 1 | 2386 | |||
AK289742.1 | - | 5 | 1707 | |||
BC047702.2 | - | 4 | 1687 | 5585128 |
