siRNA Details
Gene Symbol: PWWP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138499.3 | NP_612508.3 | 2 | 1698 | ||
XM_006717659.3 | XP_006717722.1 | 2 | 1700 | |||
GenBank mRNA | AF318365.1 | - | 1 | 1349 | ||
AK128663.1 | - | 1 | 1534 | |||
BC011630.2 | - | 1 | 1170 | |||
BC068574.1 | - | 2 | 1696 | 30344364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
