siRNA Details
Gene Symbol: ZNF675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138330.2 | NP_612203.2 | 4 | 765 | ||
XM_017026442.1 | XP_016881931.1 | 5 | 878 | |||
XM_017026443.1 | XP_016881932.1 | 5 | 1383 | |||
XM_017026444.1 | XP_016881933.1 | 4 | 762 | |||
GenBank mRNA | AB209601.1 | - | 1 | 2799 | ||
AK307845.1 | - | 4 | 693 | |||
AY044432.1 | - | 4 | 690 |
