siRNA Details
Gene Symbol: C1orf168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004303.4 | NP_001004303.3 | 18 | 2520 | ||
XM_005270584.3 | XP_005270641.1 | 14 | 2196 | |||
XM_011540898.1 | XP_011539200.1 | 19 | 2598 | |||
XM_011540899.1 | XP_011539201.1 | 17 | 2508 | |||
XM_011540904.2 | XP_011539206.1 | 16 | 1345 | |||
XM_011540906.2 | XP_011539208.1 | 14 | 1113 | |||
XM_017000544.1 | XP_016856033.1 | 19 | 2284 | |||
XM_017000545.1 | XP_016856034.1 | 19 | 2344 | |||
GenBank mRNA | AK093468.1 | - | 12 | 1031 | ||
BX648439.1 | - | 18 | 2196 |
