siRNA Details
Gene Symbol: FOXD4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012184.4 | NP_036316.1 | 1 | 1876 | ||
GenBank mRNA | AK122950.1 | - | 1 | 1876 | ||
AK125398.1 | - | 1 | 2295 | |||
AY344639.1 | - | 1 | 1788 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
