siRNA Details
Gene Symbol: SLC44A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130058.1 | NP_001123530.1 | 13 | 1079 | ||
NM_001320283.1 | NP_001307212.1 | 11 | 1040 | |||
NM_001320285.1 | NP_001307214.1 | 15 | 1239 | |||
NM_001320287.1 | NP_001307216.1 | 14 | 1172 | |||
NM_152697.4 | NP_689910.2 | 13 | 1079 | |||
XM_006710445.3 | XP_006710508.1 | 11 | 966 | |||
XM_011540986.2 | XP_011539288.1 | 12 | 1057 | |||
XM_011540987.2 | XP_011539289.1 | 14 | 1134 | |||
XM_017000607.1 | XP_016856096.1 | 12 | 1105 | |||
XM_017000608.1 | XP_016856097.1 | 12 | 1104 | |||
XM_017000609.1 | XP_016856098.1 | 15 | 2423 | |||
XM_017000610.1 | XP_016856099.1 | 15 | 1501 | |||
XM_017000611.1 | XP_016856100.1 | 13 | 1186 | |||
GenBank mRNA | AK091400.1 | - | 1 | 19 | ||
AK093170.1 | - | 13 | 1081 | |||
AK128063.1 | - | 14 | 1132 | |||
AK296924.1 | - | 12 | 1118 | |||
AK296941.1 | - | 11 | 939 | |||
AK299573.1 | - | 11 | 1040 | |||
BC028743.2 | - | 13 | 1052 | 4828797 | ||
BC034580.1 | - | 15 | 1218 | 4824861 | ||
BC051740.1 | - | 15 | 1218 | 4824908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
