siRNA Details
Gene Symbol: SLC44A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093170.1 | - | 7 | 465 | ||
AK128063.1 | - | 8 | 516 | |||
AK296924.1 | - | 6 | 502 | |||
AK296941.1 | - | 5 | 323 | |||
BC028743.2 | - | 7 | 436 | 4828797 | ||
BC034580.1 | - | 9 | 602 | 4824861 | ||
BC051740.1 | - | 9 | 602 | 4824908 | ||
BG718149.1 | - | 7 | 470 | |||
DB450780.1 | - | 7 | 464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
