siRNA Details
Gene Symbol: TRIML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173553.2 | NP_775824.2 | 4, 5 | 837 | ||
XM_011531737.2 | XP_011530039.1 | 5, 6 | 1028 | |||
XM_011531738.1 | XP_011530040.1 | 5, 6 | 1198 | |||
XM_011531739.1 | XP_011530041.1 | 3, 4 | 440 | |||
XM_017007882.1 | XP_016863371.1 | 5, 6 | 883 | |||
GenBank mRNA | AK098667.1 | - | 4, 5 | 504 | ||
BC111959.1 | - | 4, 5 | 498 | 8327427 | ||
BC111961.1 | - | 4, 5 | 498 | 8327429 |
