siRNA Details
Gene Symbol: STOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130159.2 | NP_001123631.1 | 2 | 502 | ||
NM_001130160.2 | NP_001123632.1 | 2 | 502 | |||
NM_001130161.2 | NP_001123633.1 | 2 | 502 | |||
NM_152709.4 | NP_689922.3 | 2 | 502 | |||
XM_011539454.2 | XP_011537756.1 | 2 | 233 | |||
GenBank mRNA | AK124681.1 | - | 2 | 197 | ||
AY842014.1 | - | 2 | 415 | |||
AY842015.1 | - | 2 | 415 | |||
AY842016.1 | - | 2 | 415 | |||
AY842017.1 | - | 2 | 415 | |||
BC140011.1 | - | 2 | 502 | |||
DA118241.1 | - | 2 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
