siRNA Details
Gene Symbol: STOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130161.2 | NP_001123633.1 | 3 | 1170 | ||
NM_152709.4 | NP_689922.3 | 3 | 1170 | |||
XM_011539454.2 | XP_011537756.1 | 3 | 901 | |||
GenBank mRNA | AK124681.1 | - | 3 | 865 | ||
AK127878.1 | - | 1 | 211 | |||
AY842014.1 | - | 3 | 1083 | |||
AY842017.1 | - | 3 | 1083 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
