siRNA Details
Gene Symbol: DLEU7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198989.3 | NP_945340.2 | 2 | 817 | ||
GenBank mRNA | AK126830.1 | - | 2 | 817 | ||
BC104892.1 | - | 2 | 627 | 8143895 | ||
BC112009.1 | - | 2 | 627 | 8327477 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
