siRNA Details
Gene Symbol: ERCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000400.3 | NP_000391.1 | 8 | 663 | ||
NM_001130867.1 | NP_001124339.1 | 7 | 966 | |||
XM_011526611.2 | XP_011524913.1 | 7 | 645 | |||
XM_017026467.1 | XP_016881956.1 | 7 | 559 | |||
GenBank mRNA | AL535248.3 | - | 8 | 689 | ||
BC008346.1 | - | 7 | 763 | |||
BC108255.1 | - | 8 | 626 | 6044425 | ||
BC110522.1 | - | 6 | 410 | 40034047 | ||
BC110523.1 | - | 8 | 644 | 40034048 | ||
BT006883.1 | - | 7 | 544 | |||
BX376468.2 | - | 8 | 647 | |||
X52221.1 | - | 7 | 1297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
