siRNA Details
Gene Symbol: SKA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166017.1 | NP_001159489.1 | 2, 3 | 291 | ||
NM_145061.5 | NP_659498.4 | 2, 3 | 291 | |||
XM_011534994.1 | XP_011533296.1 | 2, 3 | 267 | |||
GenBank mRNA | AF361358.1 | - | 2, 3 | 233 | ||
AK090584.1 | - | 2, 3 | 236 | |||
AK307976.1 | - | 3, 4 | 166 | |||
AK314864.1 | - | 2, 3 | 237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
