siRNA Details
Gene Symbol: SKA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145061.5 | NP_659498.4 | 8 | 1281 | ||
XM_005266288.3 | XP_005266345.1 | 7 | 1067 | |||
XM_011534994.1 | XP_011533296.1 | 8 | 1257 | |||
GenBank mRNA | AF361358.1 | - | 8 | 1223 | ||
AK314864.1 | - | 8 | 1227 | |||
BC013418.2 | - | 7 | 1165 | 3604003 | ||
BC048988.1 | - | 8 | 1252 | 5267965 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
