siRNA Details
Gene Symbol: PRSS54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080492.1 | NP_001073961.1 | 4, 5 | 666 | ||
NM_001305173.1 | NP_001292102.1 | 4, 5 | 663 | |||
GenBank mRNA | AK058068.1 | - | 4, 5 | 608 | ||
BC057843.1 | - | 4, 5 | 655 | 5296090 |
