siRNA Details
Gene Symbol: LRRC73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012974.2 | NP_001012992.1 | 4 | 1157 | ||
NM_001271882.1 | NP_001258811.1 | 4 | 465 | |||
GenBank mRNA | AK126786.1 | - | 3 | 377 | ||
BC107130.2 | - | 4 | 365 | |||
BC108661.1 | - | 4 | 1157 | 5240367 | ||
HY024930.1 | - | 4 | 465 |
